Assay Details
Gene Symbol: IER5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005540158.2 | XP_005540215.1 | 1 | 1774 | 57 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005540158.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
