Assay Details
Gene Symbol: PLPP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005556904.2 | XP_005556961.1 | 1 - 2 | 508 | 134 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
