Assay Details
Gene Symbol: TOR1AIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005540140.2 | XP_005540197.1 | 6 - 7 | 1456 | 90 | |
XM_005540144.2 | XP_005540201.1 | 4 - 5 | 1433 | 90 | ||
XM_015446340.1 | XP_015301826.1 | 5 - 6 | 1040 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
