This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mf02868691_m1
See other ALMS1 GE Assays ›
Gene Symbol:
ALMS1
Gene Name:
ALMS1 centrosome and basal body associated protein
Gene Aliases:
-
Chromosome Location:
Chr.13: 36241011 - 36463180 on Build M_fas_5.0
Species:
Macaca fascicularis
Species Specific ID (Flybase ID):
-
Assay ID Mf02868691_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ALMS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_015433535.1 XP_015289021.1 1 - 2 353 108
XM_015433536.1 XP_015289022.1 1 - 2 353 108
XM_015433537.1 XP_015289023.1 1 - 2 353 108
XM_015433539.1 XP_015289025.1 1 - 2 353 108
XM_015433540.1 XP_015289026.1 1 - 2 353 108

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Recommended Assay Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7db4fb6694-gx648:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header