This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mf02858459_m1
See other GIPC1 GE Assays ›
Gene Symbol:
GIPC1
Gene Name:
GIPC PDZ domain containing family member 1
Gene Aliases:
-
Chromosome Location:
Chr.19: 14864669 - 14885172 on Build M_fas_5.0
Species:
Macaca fascicularis
Species Specific ID (Flybase ID):
-
Assay ID Mf02858459_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GIPC1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005588250.2 XP_005588307.1 4 - 5 557 100
XM_015440381.1 XP_015295867.1 4 - 5 483 100
XM_015440382.1 XP_015295868.1 4 - 5 665 100
XM_015440383.1 XP_015295869.1 4 - 5 329 100
XM_015440384.1 XP_015295870.1 3 - 4 248 100

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_015440383.1, XM_015440384.1 transcripts.

Set Membership:

Amplicon length between 86 and 100 3' Most Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

scaffold/adaptor protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7db4fb6694-gx648:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header