Assay Details
Gene Symbol: ZCCHC7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005581281.2 | XP_005581338.1 | 7 - 8 | 1248 | 124 | |
XM_005581282.2 | XP_005581339.1 | 7 - 8 | 1252 | 124 | ||
XM_005581283.2 | XP_005581340.1 | 7 - 8 | 1620 | 124 | ||
XM_005581284.2 | XP_005581341.1 | 7 - 8 | 1876 | 124 | ||
XM_005581285.2 | XP_005581342.1 | 7 - 8 | 1284 | 124 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
