Assay Details
Gene Symbol: BAX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005589832.2 | XP_005589889.1 | 2 - 3 | 184 | 129 | |
XM_015441196.1 | XP_015296682.1 | 3 - 4 | 796 | 129 | ||
XM_015441197.1 | XP_015296683.1 | 2 - 3 | 111 | 129 | ||
XM_015441201.1 | XP_015296687.1 | 2 - 3 | 187 | 129 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015441197.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
