This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mf02836693_m1
See other MDH1B GE Assays ›
Gene Symbol:
MDH1B
Gene Name:
malate dehydrogenase 1B
Gene Aliases:
-
Chromosome Location:
Chr.12: 96328674 - 96368281 on Build M_fas_5.0
Species:
Macaca fascicularis
Species Specific ID (Flybase ID):
-
Assay ID Mf02836693_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MDH1B
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005574066.2 XP_005574123.1 8 - 9 1657 134
XM_005574067.2 XP_005574124.1 8 - 9 1615 134
XM_005574070.2 XP_005574127.1 8 - 9 1615 134
XM_015432651.1 XP_015288137.1 8 - 9 1465 134
XM_015432652.1 XP_015288138.1 8 - 9 1465 134
XM_015432653.1 XP_015288139.1 8 - 9 1423 134
XM_015432654.1 XP_015288140.1 8 - 9 1465 134
XM_015432655.1 XP_015288141.1 8 - 9 1465 134
XM_015432656.1 XP_015288142.1 8 - 9 1465 134
XM_015432657.1 XP_015288143.1 8 - 9 1615 134
XM_015432658.1 XP_015288144.1 7 - 8 1362 134

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

dehydrogenase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0