Assay Details
Gene Symbol: RGS7BP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005556997.2 | XP_005557054.1 | 3 - 4 | 1433 | 147 | |
XM_005556998.2 | XP_005557055.1 | 3 - 4 | 564 | 147 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
