This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mf02810123_g1
See other RQCD1 GE Assays ›
Gene Symbol:
RQCD1
Gene Name:
CCR4-NOT transcription complex subunit 9
Gene Aliases:
CNOT9, RCD1, Rcd-1
Chromosome Location:
Chr.12: 108239340 - 108270750 on Build M_fas_5.0
Species:
Macaca fascicularis
Species Specific ID (Flybase ID):
-
Assay ID Mf02810123_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RQCD1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001283775.1 NP_001270704.1 2 - 3 350 76
XM_005574286.2 XP_005574343.1 2 - 3 615 76
XM_015432080.1 XP_015287566.1 1 - 2 530 76
XM_015432081.1 XP_015287567.1 1 - 2 531 76
XM_015432082.1 XP_015287568.1 1 - 2 531 76
XM_015432083.1 XP_015287569.1 2 - 3 378 76
GenBank mRNA AB179421.1 - 2 - 3 350 76

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0