Assay Details
Gene Symbol: RACGAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005570801.2 | XP_005570858.1 | 12 - 13 | 1463 | 149 | |
XM_005570802.1 | XP_005570859.1 | 12 - 13 | 1553 | 149 | ||
XM_005570803.2 | XP_005570860.1 | 12 - 13 | 1456 | 149 | ||
XM_005570804.2 | XP_005570861.1 | 12 - 13 | 1390 | 149 | ||
XM_005570805.2 | XP_005570862.1 | 12 - 13 | 1478 | 149 | ||
XM_005570806.2 | XP_005570863.1 | 12 - 13 | 1363 | 149 | ||
XM_005570807.2 | XP_005570864.1 | 11 - 12 | 1276 | 149 | ||
XM_015430898.1 | XP_015286384.1 | 12 - 13 | 1362 | 149 | ||
XM_015430899.1 | XP_015286385.1 | 11 - 12 | 1374 | 149 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
