This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mf02808382_m1
See other NFYB GE Assays ›
Gene Symbol:
NFYB
Gene Name:
nuclear transcription factor Y subunit beta
Gene Aliases:
-
Chromosome Location:
Chr.11: 107807962 - 107828419 on Build M_fas_5.0
Species:
Macaca fascicularis
Species Specific ID (Flybase ID):
-
Assay ID Mf02808382_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NFYB
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005572068.2 XP_005572125.1 5 - 6 466 87
XM_005572069.2 XP_005572126.1 4 - 5 2194 87
XM_005572070.2 XP_005572127.1 5 - 6 463 87
XM_005572071.2 XP_005572128.1 4 - 5 381 87
XM_015431466.1 XP_015286952.1 3 - 4 2108 87
XM_015431467.1 XP_015286953.1 4 - 5 412 87

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

DNA-binding transcription factor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0