This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mf02805457_m1
See other STARD10 GE Assays ›
Gene Symbol:
STARD10
Gene Name:
StAR related lipid transfer domain containing 10
Gene Aliases:
-
Chromosome Location:
Chr.14: 67936995 - 67978930 on Build M_fas_5.0
Species:
Macaca fascicularis
Species Specific ID (Flybase ID):
-
Assay ID Mf02805457_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: STARD10
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005579008.2 XP_005579065.1 5 - 6 912 90
XM_005579010.2 XP_005579067.1 5 - 6 842 90
XM_005579011.2 XP_005579068.1 6 - 7 2150 90
XM_005579012.2 XP_005579069.1 5 - 6 709 90
XM_015435452.1 XP_015290938.1 5 - 6 2095 90
XM_015435453.1 XP_015290939.1 7 - 8 1614 90
XM_015435454.1 XP_015290940.1 5 - 6 778 90
GenBank mRNA AB168575.1 - 6 - 7 90

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Recommended Assay Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0