This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mf02802855_m1
See other PPP2R5A GE Assays ›
Gene Symbol:
PPP2R5A
Gene Name:
protein phosphatase 2 regulatory subunit B'alpha
Gene Aliases:
-
Chromosome Location:
Chr.1: 70878493 - 70954152 on Build M_fas_5.0
Species:
Macaca fascicularis
Species Specific ID (Flybase ID):
-
Assay ID Mf02802855_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PPP2R5A
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005540775.2 XP_005540832.1 7 - 8 1452 102
XM_005540776.2 XP_005540833.1 7 - 8 1931 102
XM_005540777.2 XP_005540834.1 7 - 8 2008 102
XM_005540778.2 XP_005540835.1 7 - 8 2061 102
XM_015449817.1 XP_015305303.1 7 - 8 1691 102
XM_015449819.1 XP_015305305.1 7 - 8 715 102
GenBank mRNA AB173675.1 - 7 - 8 1030 102

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

protein phosphatase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56ffd697dc-88d4p:80/100.66.130.94:80.
git-commit: 082a6d415be982b1f92c35e55dd71609d4576d3e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0