This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mf02801192_m1
See other MTFR1L GE Assays ›
Gene Symbol:
MTFR1L
Gene Name:
mitochondrial fission regulator 1 like
Gene Aliases:
-
Chromosome Location:
Chr.1: 202529906 - 202543468 on Build M_fas_5.0
Species:
Macaca fascicularis
Species Specific ID (Flybase ID):
-
Assay ID Mf02801192_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MTFR1L
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005544382.1 XP_005544439.1 6 - 7 971 94
XM_005544383.2 XP_005544440.1 6 - 7 1079 94
XM_005544384.2 XP_005544441.1 6 - 7 1251 94
XM_015440565.1 XP_015296051.1 6 - 7 971 94
XM_015440568.1 XP_015296054.1 6 - 7 1169 94
XM_015440572.1 XP_015296058.1 6 - 7 935 94
XM_015440580.1 XP_015296066.1 6 - 7 862 94

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Recommended Assay Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0