This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mf02799075_m1
See other GOLGA7 GE Assays ›
Gene Symbol:
GOLGA7
Gene Name:
golgin A7
Gene Aliases:
-
Chromosome Location:
Chr.8: 42451613 - 42471546 on Build M_fas_5.0
Species:
Macaca fascicularis
Species Specific ID (Flybase ID):
-
Assay ID Mf02799075_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GOLGA7
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001285046.1 NP_001271975.1 1 - 2 295 118
XM_015454357.1 XP_015309843.1 2 - 3 279 118
XM_015454358.1 XP_015309844.1 2 - 3 302 118
XM_015454359.1 XP_015309845.1 2 - 3 192 118
XM_015454360.1 XP_015309846.1 2 - 3 279 118
GenBank mRNA AB173135.1 - 1 - 2 295 118

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Recommended Assay Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0