Assay Details
Gene Symbol: DRG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001319407.1 | NP_001306336.1 | 1 - 2 | 170 | 54 | |
XM_005583056.2 | XP_005583113.1 | 1 - 2 | 204 | 54 | ||
XM_015437729.1 | XP_015293215.1 | 1 - 2 | 204 | 54 | ||
XM_015437730.1 | XP_015293216.1 | 1 - 2 | 204 | 54 | ||
XM_015437731.1 | XP_015293217.1 | 1 - 2 | 222 | 54 | ||
GenBank mRNA | AB170580.1 | - | 1 - 2 | 170 | 54 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015437731.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
