This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mf02787898_m1
See other OPRM1 GE Assays ›
Gene Symbol:
OPRM1
Gene Name:
opioid receptor mu 1
Gene Aliases:
M-OR-1, MOR-1, MOR1
Chromosome Location:
Chr.4: 60331340 - 60455610 on Build M_fas_5.0
Species:
Macaca fascicularis
Species Specific ID (Flybase ID):
-
Assay ID Mf02787898_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: OPRM1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001287630.1 NP_001274559.1 2 - 3 723 95
XM_005552171.2 XP_005552228.1 2 - 3 707 95
XM_005552172.2 XP_005552229.1 2 - 3 707 95
XM_005552177.2 XP_005552234.1 2 - 3 884 95
XM_015448824.1 XP_015304310.1 3 - 4 1312 95
GenBank mRNA AY038989.1 - 2 - 3 723 95

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

G-protein coupled receptor transmembrane signal receptor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0