This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mf01118529_m1
See other TUT4 GE Assays ›
Gene Symbol:
TUT4
Gene Name:
terminal uridylyl transferase 4
Gene Aliases:
ZCCHC11
Chromosome Location:
Chr.1: 174957064 - 175089581 on Build M_fas_5.0
Species:
Macaca fascicularis
Species Specific ID (Flybase ID):
-
Assay ID Mf01118529_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TUT4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005543357.2 XP_005543414.1 19 - 20 3568 118
XM_005543358.2 XP_005543415.1 19 - 20 3568 118
XM_005543359.2 XP_005543416.1 19 - 20 3568 118
XM_005543360.2 XP_005543417.1 19 - 20 3571 118
XM_005543361.2 XP_005543418.1 19 - 20 3571 118
XM_005543362.2 XP_005543419.1 19 - 20 3568 118
XM_005543363.2 XP_005543420.1 19 - 20 3487 118
XM_005543364.2 XP_005543421.1 18 - 19 3481 118
XM_005543365.2 XP_005543422.1 18 - 19 3274 118
XM_015435819.1 XP_015291305.1 20 - 21 3621 118
XM_015435833.1 XP_015291319.1 18 - 19 3355 118
XM_015435838.1 XP_015291324.1 17 - 18 3235 118

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

nucleotidyltransferase transferase metabolite interconversion enzyme

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0