Assay Details
Gene Symbol: RQCD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283775.1 | NP_001270704.1 | 5 - 6 | 692 | 87 | |
XM_005574286.2 | XP_005574343.1 | 5 - 6 | 957 | 87 | ||
XM_015432080.1 | XP_015287566.1 | 4 - 5 | 872 | 87 | ||
XM_015432081.1 | XP_015287567.1 | 4 - 5 | 873 | 87 | ||
XM_015432082.1 | XP_015287568.1 | 4 - 5 | 873 | 87 | ||
XM_015432083.1 | XP_015287569.1 | 5 - 6 | 720 | 87 | ||
GenBank mRNA | AB179421.1 | - | 5 - 6 | 692 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
