Assay Details
Gene Symbol: CLDND1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005548338.2 | XP_005548395.2 | 5 - 6 | 826 | 97 | |
XM_005548339.2 | XP_005548396.1 | 4 - 5 | 785 | 97 | ||
XM_005548340.2 | XP_005548397.1 | 5 - 6 | 824 | 97 | ||
XM_005548341.2 | XP_005548398.1 | 5 - 6 | 765 | 97 | ||
XM_005548342.2 | XP_005548399.1 | 5 - 6 | 736 | 97 | ||
XM_005548343.2 | XP_005548400.1 | 5 - 6 | 689 | 97 | ||
XM_005548344.2 | XP_005548401.1 | 4 - 5 | 1088 | 97 | ||
XM_015446558.1 | XP_015302044.1 | 5 - 6 | 920 | 97 | ||
XM_015446559.1 | XP_015302045.1 | 5 - 6 | 915 | 97 | ||
XM_015446560.1 | XP_015302046.1 | 5 - 6 | 706 | 97 | ||
XM_015446561.1 | XP_015302047.1 | 5 - 6 | 659 | 97 | ||
XM_015446562.1 | XP_015302048.1 | 5 - 6 | 665 | 97 | ||
GenBank mRNA | AB169751.1 | - | 5 - 6 | 732 | 97 | |
AB174462.1 | - | 5 - 6 | 685 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
