This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mf00952047_m1
See other GPR132 GE Assays ›
Gene Symbol:
GPR132
Gene Name:
G protein-coupled receptor 132
Gene Aliases:
-
Chromosome Location:
Chr.7: 170421397 - 170438305 on Build M_fas_5.0
Species:
Macaca fascicularis
Species Specific ID (Flybase ID):
-
Assay ID Mf00952047_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GPR132
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005562369.2 XP_005562426.1 1 - 2 902 116
XM_015454265.1 XP_015309751.1 1 - 2 643 116
XM_015454266.1 XP_015309752.1 1 - 2 643 116

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_005562369.2, XM_015454265.1, XM_015454266.1 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Probe spans exons

Panther Classification:

Molecular Function -

G-protein coupled receptor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0