This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mf00897590_m1
See other SLA GE Assays ›
Gene Symbol:
SLA
Gene Name:
Src like adaptor
Gene Aliases:
-
Chromosome Location:
Chr.8: 134411106 - 134478351 on Build M_fas_5.0
Species:
Macaca fascicularis
Species Specific ID (Flybase ID):
-
Assay ID Mf00897590_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SLA
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001287302.1 NP_001274231.1 4 - 5 469 71
XM_005564113.2 XP_005564170.1 3 - 4 452 71
XM_005564114.2 XP_005564171.1 8 - 9 2471 71
XM_015454794.1 XP_015310280.1 6 - 7 810 71
XM_015454795.1 XP_015310281.1 5 - 6 820 71
XM_015454797.1 XP_015310283.1 8 - 9 1510 71
XM_015454798.1 XP_015310284.1 8 - 9 1307 71
GenBank mRNA AB172409.1 - 4 - 5 469 71

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

kinase modulator

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-867854cbc7-vwcfx:80/100.66.130.94:80.
git-commit: 25ccc7b1e9fd8ebf0690872e1fef11e169411d8b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0