This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mf00469699_mH
See other ZFAND6 GE Assays ›
Gene Symbol:
ZFAND6
Gene Name:
zinc finger AN1-type containing 6
Gene Aliases:
-
Chromosome Location:
Chr.7: 58519297 - 58602128 on Build M_fas_5.0
Species:
Macaca fascicularis
Species Specific ID (Flybase ID):
-
Assay ID Mf00469699_mH
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ZFAND6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001283570.1 NP_001270499.1 5 - 6 624 65
XM_015452445.1 XP_015307931.1 6 - 7 900 65
XM_015452446.1 XP_015307932.1 6 - 7 3363 65
XM_015452447.1 XP_015307933.1 6 - 7 877 65
XM_015452448.1 XP_015307934.1 5 - 6 731 65
XM_015452449.1 XP_015307935.1 5 - 6 3201 65
XM_015452450.1 XP_015307936.1 5 - 6 705 65
XM_015452451.1 XP_015307937.1 5 - 6 512 65
GenBank mRNA AB170979.1 - 5 - 6 628 65
AB171334.1 - 5 - 6 618 65
AB174204.1 - 5 - 6 624 65

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Recommended Assay Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header