This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07291855_s1
See other RANGRF GE Assays ›
Gene Symbol:
RANGRF
Gene Name:
RAN guanine nucleotide release factor
Gene Aliases:
HSPC165, HSPC236, MOG1, RANGNRF
Chromosome Location:
Chr.17: 8287150 - 8290091 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs07291855_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RANGRF
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001177801.1 NP_001171272.1 4 633 70
NM_001177802.1 NP_001171273.1 3 721 70
GenBank mRNA AF265205.1 - 4 613 70
AK290399.1 - 4 632 70
BC006486.1 - 3 687 3536119 70
BC012552.1 - 3 694 4134442 70
BC100017.1 - 4 614 6156156 70

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Gene Ontology Categories:

Function(s) Process(es)

regulation of heart rate
regulation of membrane depolarization
ER to Golgi vesicle-mediated transport
protein exit from endoplasmic reticulum
regulation of membrane potential
positive regulation of GTPase activity
positive regulation of establishment of protein localization to plasma membrane
regulation of membrane depolarization during cardiac muscle cell action potential
regulation of sodium ion transmembrane transport
positive regulation of protein localization to cell surface
regulation of sodium ion transmembrane transporter activity
guanyl-nucleotide exchange factor activity
Ran guanyl-nucleotide exchange factor activity
Ran GTPase binding
protein transporter activity
sodium channel regulator activity
ion channel binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0