This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07291851_s1
See other RANGRF GE Assays ›
Gene Symbol:
RANGRF
Gene Name:
RAN guanine nucleotide release factor
Gene Aliases:
HSPC165, HSPC236, MOG1, RANGNRF
Chromosome Location:
Chr.17: 8287150 - 8290091 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs07291851_s1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RANGRF
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001177801.1 NP_001171272.1 4 716 77
NM_001177802.1 NP_001171273.1 3 804 77
GenBank mRNA AF265205.1 - 4 696 77
AK290399.1 - 4 715 77
BC006486.1 - 3 770 3536119 77
BC012552.1 - 3 777 4134442 77
BC100017.1 - 4 697 6156156 77

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Gene Ontology Categories:

Function(s) Process(es)

regulation of heart rate
regulation of membrane depolarization
ER to Golgi vesicle-mediated transport
protein exit from endoplasmic reticulum
regulation of membrane potential
positive regulation of GTPase activity
positive regulation of establishment of protein localization to plasma membrane
regulation of membrane depolarization during cardiac muscle cell action potential
regulation of sodium ion transmembrane transport
positive regulation of protein localization to cell surface
regulation of sodium ion transmembrane transporter activity
guanyl-nucleotide exchange factor activity
Ran guanyl-nucleotide exchange factor activity
Ran GTPase binding
protein transporter activity
sodium channel regulator activity
ion channel binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0