This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07291796_m1
See other CARD16 GE Assays ›
Gene Symbol:
CARD16
Gene Name:
caspase recruitment domain family member 16
Gene Aliases:
COP, COP1, PSEUDO-ICE
Chromosome Location:
Chr.11: 105040218 - 105045416 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs07291796_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CARD16
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001017534.1 NP_001017534.1 2 - 3 299 98
XM_011542583.2 XP_011540885.1 2 - 3 491 98
GenBank mRNA AY885669.1 - 2 - 3 290 98

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Homo sapiens Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene CARD16

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of endopeptidase activity
regulation of apoptotic process
cysteine-type endopeptidase inhibitor activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0