This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07291685_m1
See other SH3GL1 GE Assays ›
Gene Symbol:
SH3GL1
Gene Name:
SH3 domain containing GRB2 like 1, endophilin A2
Gene Aliases:
CNSA1, EEN, SH3D2B, SH3P8
Chromosome Location:
Chr.19: 4360367 - 4400568 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs07291685_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SH3GL1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001199944.1 NP_001186873.1 4 - 5 288 100
GenBank mRNA AK097616.1 - 4 - 5 288 100

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Homo sapiens Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

endocytosis
signal transduction
central nervous system development
cell-cell adhesion
protein binding
lipid binding
phosphatase binding
identical protein binding
GTPase binding
cadherin binding involved in cell-cell adhesion

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0