This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07291513_m1
See other NOL3 GE Assays ›
Gene Symbol:
NOL3
Gene Name:
nucleolar protein 3
Gene Aliases:
ARC, FCM, MYP, NOP, NOP30
Chromosome Location:
Chr.16: 67170497 - 67175737 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs07291513_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NOL3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001276307.1 NP_001263236.1 3 - 4 223 76
GenBank mRNA CB959673.1 - 3 - 4 250 76

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001276307.1 transcripts.

Set Membership:

Amplicon length between 71 and 85 Homo sapiens Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

response to hypoxia
blood vessel remodeling
response to ischemia
mRNA splice site selection
RNA splicing
cardiac muscle cell apoptotic process
negative regulation of cardiac muscle cell apoptotic process
negative regulation of tumor necrosis factor-mediated signaling pathway
negative regulation of muscle atrophy
release of sequestered calcium ion into cytosol by sarcoplasmic reticulum
response to injury involved in regulation of muscle adaptation
regulation of NF-kappaB import into nucleus
negative regulation of apoptotic process
protein oligomerization
negative regulation of necrotic cell death
negative regulation of release of cytochrome c from mitochondria
intrinsic apoptotic signaling pathway
negative regulation of mitochondrial membrane permeability involved in apoptotic process
negative regulation of oxidative stress-induced intrinsic apoptotic signaling pathway
inhibition of cysteine-type endopeptidase activity involved in apoptotic process
negative regulation of extrinsic apoptotic signaling pathway
RNA binding
death receptor binding
calcium ion binding
protein binding
death effector domain binding
identical protein binding
cysteine-type endopeptidase inhibitor activity involved in apoptotic process
caspase binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-867854cbc7-vwcfx:80/100.66.130.94:80.
git-commit: 25ccc7b1e9fd8ebf0690872e1fef11e169411d8b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0