This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07291479_m1
See other SPINK2 GE Assays ›
Gene Symbol:
SPINK2
Gene Name:
serine peptidase inhibitor, Kazal type 2
Gene Aliases:
HUSI-II
Chromosome Location:
Chr.4: 56809860 - 56822210 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs07291479_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SPINK2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001271719.1 NP_001258648.1 1 - 3 289 103
GenBank mRNA BI520020.1 - 1 - 3 218 103

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Homo sapiens

Panther Classification:

Molecular Function -

protease inhibitor

Gene Ontology Categories:

Function(s) Process(es)

regulation of acrosome reaction
negative regulation of serine-type endopeptidase activity
endopeptidase inhibitor activity
serine-type endopeptidase inhibitor activity
protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0