This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07291414_g1
See other EPGN GE Assays ›
Gene Symbol:
EPGN
Gene Name:
epithelial mitogen
Gene Aliases:
ALGV3072, EPG, PRO9904
Chromosome Location:
Chr.4: 74307571 - 74317298 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs07291414_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: EPGN
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001270992.1 NP_001257921.1 3 - 4 318 71
NM_001270993.1 NP_001257922.1 3 - 4 291 71
NR_073117.1 2 - 3 228 71
GenBank mRNA DQ235265.1 - 3 - 4 227 71
DQ235266.1 - 2 - 3 164 71
DQ235267.1 - 3 - 4 254 71

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

growth factor

Gene Ontology Categories:

Function(s) Process(es)

MAPK cascade
activation of MAPK activity
angiogenesis
positive regulation of cell proliferation
positive regulation of MAP kinase activity
positive regulation of epidermal growth factor-activated receptor activity
positive regulation of mitotic nuclear division
positive regulation of epithelial cell proliferation
epidermal growth factor receptor binding
growth factor activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0