This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07291387_m1
See other PKIB GE Assays ›
Gene Symbol:
PKIB
Gene Name:
protein kinase (cAMP-dependent, catalytic) inhibitor beta
Gene Aliases:
PRKACN2
Chromosome Location:
Chr.6: 122471917 - 122726373 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs07291387_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PKIB
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001270394.1 NP_001257323.1 6 - 7 507 103
NM_001270395.1 NP_001257324.1 3 - 4 168 103
XM_011535930.2 XP_011534232.1 4 - 5 559 103
XM_011535931.2 XP_011534233.1 5 - 6 330 103
XM_011535932.2 XP_011534234.1 4 - 5 304 103
XM_011535933.2 XP_011534235.1 4 - 5 265 103
XM_011535934.2 XP_011534236.1 2 - 3 136 103
GenBank mRNA AK074397.1 - 3 - 4 167 103
BP262756.1 - 3 - 4 168 103

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001270394.1, NM_001270395.1, XM_011535930.2, XM_011535931.2, XM_011535932.2, XM_011535933.2, XM_011535934.2 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

kinase inhibitor

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of telomere maintenance via telomerase
positive regulation of telomerase activity
positive regulation of telomere capping
negative regulation of cAMP-dependent protein kinase activity
cAMP-dependent protein kinase inhibitor activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0