This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07291134_m1
See other ZFAND6 GE Assays ›
Gene Symbol:
ZFAND6
Gene Name:
zinc finger AN1-type containing 6
Gene Aliases:
AWP1, ZA20D3, ZFAND5B
Chromosome Location:
Chr.15: 80059568 - 80138393 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs07291134_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ZFAND6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001242917.1 NP_001229846.1 2 - 3 290 98
GenBank mRNA AF061739.1 - 2 - 3 231 98

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AF061739.1, NM_001242917.1 transcripts.

Set Membership:

Amplicon length between 86 and 100 3' Most Homo sapiens Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

protein targeting to peroxisome
apoptotic process
negative regulation of apoptotic process
regulation of I-kappaB kinase/NF-kappaB signaling
cellular response to tumor necrosis factor
molecular_function
DNA binding
protein binding
zinc ion binding
polyubiquitin binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0