This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07290574_g1
See other NOL3 GE Assays ›
Gene Symbol:
NOL3
Gene Name:
nucleolar protein 3
Gene Aliases:
ARC, FCM, MYP, NOP, NOP30
Chromosome Location:
Chr.16: 67170497 - 67175737 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs07290574_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NOL3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001185057.2 NP_001171986.1 2 - 3 484 58
XM_005256217.2 XP_005256274.1 2 - 3 1121 58
XM_011523424.2 XP_011521726.1 3 - 4 964 58
GenBank mRNA AF064599.1 - 2 - 3 335 58

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Homo sapiens Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

response to hypoxia
blood vessel remodeling
response to ischemia
mRNA splice site selection
RNA splicing
cardiac muscle cell apoptotic process
negative regulation of cardiac muscle cell apoptotic process
negative regulation of tumor necrosis factor-mediated signaling pathway
negative regulation of muscle atrophy
release of sequestered calcium ion into cytosol by sarcoplasmic reticulum
response to injury involved in regulation of muscle adaptation
regulation of NF-kappaB import into nucleus
negative regulation of apoptotic process
protein oligomerization
negative regulation of necrotic cell death
negative regulation of release of cytochrome c from mitochondria
intrinsic apoptotic signaling pathway
negative regulation of mitochondrial membrane permeability involved in apoptotic process
negative regulation of oxidative stress-induced intrinsic apoptotic signaling pathway
inhibition of cysteine-type endopeptidase activity involved in apoptotic process
negative regulation of extrinsic apoptotic signaling pathway
RNA binding
death receptor binding
calcium ion binding
protein binding
death effector domain binding
identical protein binding
cysteine-type endopeptidase inhibitor activity involved in apoptotic process
caspase binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0