This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07290545_g1
See other GOLGA7 GE Assays ›
Gene Symbol:
GOLGA7
Gene Name:
golgin A7
Gene Aliases:
GCP16, GOLGA3AP1, GOLGA7A, HSPC041
Chromosome Location:
Chr.8: 41490562 - 41510980 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs07290545_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GOLGA7
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001002296.1 NP_001002296.1 2 - 3 469 98
NM_001174124.1 NP_001167595.1 3 - 4 346 98
NM_016099.2 NP_057183.2 3 - 4 346 98
GenBank mRNA AB104615.1 - 3 - 4 326 98
AF068291.1 - 2 - 3 300 98
AF125102.1 - 3 - 4 310 98
AK026613.1 - 3 - 4 326 98
AL695304.1 - 3 - 4 355 98
BC012032.1 - 2 - 3 469 4707385 98

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene GOLGA7

Gene Ontology Categories:

Function(s) Process(es)

protein targeting to membrane
peptidyl-L-cysteine S-palmitoylation
Golgi to plasma membrane protein transport
protein stabilization
protein binding
protein-cysteine S-palmitoyltransferase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header