This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07289737_m1
See other GDPD4 GE Assays ›
Gene Symbol:
GDPD4
Gene Name:
glycerophosphodiester phosphodiesterase domain containing 4
Gene Aliases:
GDE6
Chromosome Location:
Chr.11: 77216558 - 77301699 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs07289737_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GDPD4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_182833.1 NP_878253.1 10 - 11 1118 91
XM_011544826.1 XP_011543128.1 10 - 11 2849 91
XM_011544833.1 XP_011543135.1 10 - 11 2849 91
XM_011544834.1 XP_011543136.1 11 - 12 1344 91
XM_017017348.1 XP_016872837.1 11 - 12 1288 91
XM_017017349.1 XP_016872838.1 10 - 11 1205 91
XM_017017350.1 XP_016872839.1 10 - 11 2720 91
XM_017017351.1 XP_016872840.1 10 - 11 1076 91
GenBank mRNA AY313782.1 - 10 - 11 1118 91
AY326450.1 - 10 - 11 1118 91

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene GDPD4

Panther Classification:

Molecular Function -

phosphodiesterase

Gene Ontology Categories:

Function(s) Process(es)

lipid metabolic process
glycerophosphodiester phosphodiesterase activity
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0