This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07289187_g1
See other NPPC GE Assays ›
Gene Symbol:
NPPC
Gene Name:
natriuretic peptide C
Gene Aliases:
CNP, CNP2
Chromosome Location:
Chr.2: 231922094 - 231926403 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs07289187_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NPPC
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_024409.3 NP_077720.1 2 - 3 558 76
GenBank mRNA DR003533.1 - 2 - 3 566 76
JF420840.1 - 1 - 2 209 76

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 3' Most Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene NPPC

Panther Classification:

Molecular Function -

peptide hormone

Gene Ontology Categories:

Function(s) Process(es)

ossification
response to hypoxia
growth plate cartilage chondrocyte differentiation
growth plate cartilage chondrocyte proliferation
cGMP biosynthetic process
receptor guanylyl cyclase signaling pathway
negative regulation of cell proliferation
post-embryonic development
regulation of cAMP metabolic process
regulation of cGMP metabolic process
positive regulation of cGMP biosynthetic process
regulation of multicellular organism growth
response to drug
single organism reproductive process
response to ethanol
positive regulation of osteoblast differentiation
positive regulation of vasodilation
regulation of smooth muscle cell proliferation
negative regulation of DNA metabolic process
negative regulation of meiotic cell cycle
negative regulation of oocyte maturation
regulation of cardiac conduction
receptor binding
hormone activity
protein homodimerization activity
peptide hormone receptor binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0