This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07289091_m1
See other CABP5 GE Assays ›
Gene Symbol:
CABP5
Gene Name:
calcium binding protein 5
Gene Aliases:
CABP3
Chromosome Location:
Chr.19: 48029383 - 48048342 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs07289091_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CABP5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_019855.4 NP_062829.1 2 - 3 228 68
XM_017026992.1 XP_016882481.1 4 - 5 429 68
GenBank mRNA AF169159.1 - 2 - 3 221 68
BC126133.1 - 2 - 3 136 8991849 68
BC126135.1 - 2 - 3 136 8991851 68

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene CABP5

Gene Ontology Categories:

Function(s) Process(es)

signal transduction
calcium ion binding
protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0