This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07288847_m1
See other DYNLL1 GE Assays ›
Gene Symbol:
DYNLL1
Gene Name:
dynein light chain LC8-type 1
Gene Aliases:
DLC1, DLC8, DNCL1, DNCLC1, LC8, LC8a, PIN, hdlc1
Chromosome Location:
Chr.12: 120469857 - 120498495 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs07288847_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: DYNLL1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001037494.1 NP_001032583.1 1 - 2 246 98
GenBank mRNA BG474448.1 - 1 - 2 97
CB992195.1 - 1 - 2 273 98

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001037494.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene DYNLL1

Panther Classification:

Molecular Function -

microtubule or microtubule-binding cytoskeletal protein

Gene Ontology Categories:

Function(s) Process(es)

G2/M transition of mitotic cell cycle
transcription, DNA-templated
regulation of transcription, DNA-templated
ER to Golgi vesicle-mediated transport
apoptotic process
female gamete generation
anatomical structure morphogenesis
viral process
macroautophagy
antigen processing and presentation of exogenous peptide antigen via MHC class II
substantia nigra development
actin cytoskeleton organization
intraciliary retrograde transport
negative regulation of phosphorylation
cilium morphogenesis
positive regulation of protein insertion into mitochondrial membrane involved in apoptotic signaling pathway
motor activity
protein binding
protein C-terminus binding
protein domain specific binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0