This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07288839_m1
See other RDM1 GE Assays ›
Gene Symbol:
RDM1
Gene Name:
RAD52 motif containing 1
Gene Aliases:
RAD52B
Chromosome Location:
Chr.17: 35918080 - 35931821 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs07288839_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RDM1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001034836.1 NP_001030008.1 3 - 4 445 105
NM_001163121.1 NP_001156593.1 3 - 4 445 105
NM_001163122.1 NP_001156594.1 2 - 3 344 105
NM_001163130.1 NP_001156602.1 2 - 3 344 105
NM_145654.3 NP_663629.1 3 - 4 445 105
XM_011524509.2 XP_011522811.1 3 - 4 1490 105
GenBank mRNA AB080728.1 - 3 - 4 423 105
AK302782.1 - 2 - 3 798 105
BC127190.1 - 3 - 4 436 40036784 105
BM559857.1 - 3 - 4 422 105
EF488475.1 - 3 - 4 396 105
EF488479.1 - 2 - 3 327 105
EF488481.1 - 2 - 3 327 105

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene RDM1

Gene Ontology Categories:

Process(es)

nucleotide binding
DNA binding
RNA binding
protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0