This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07288629_m1
See other LGALS3 GE Assays ›
Gene Symbol:
LGALS3
Gene Name:
lectin, galactoside binding soluble 3
Gene Aliases:
CBP35, GAL3, GALBP, GALIG, L31, LGALS2, MAC2
Chromosome Location:
Chr.14: 55129217 - 55145430 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs07288629_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LGALS3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_002306.3 NP_002297.2 5 - 6 672 129
NR_003225.2 3 - 4 1628 129
XM_011536759.2 XP_011535061.2 6 - 7 662 129
GenBank mRNA AB006780.1 - 5 - 6 637 129
AB209391.1 - 4 - 5 4715 129
AF266280.1 - 3 - 4 955 129
AK314929.1 - 5 - 6 671 129
BC001120.2 - 5 - 6 608 3050135 129
BC053667.1 - 5 - 6 623 6148401 129
BC068068.1 - 5 - 6 618 6428428 129
BU681070.1 - 1 - 2 346 129
BX161455.1 - 3 - 4 1423 129
BX641090.1 - 3 - 4 1629 129
CD518618.1 - 5 - 6 672 129
CR456897.1 - 4 - 5 584 129
CR542097.1 - 4 - 5 584 129
M35368.1 - 5 - 6 606 129
M36682.1 - 5 - 6 617 129
M57710.1 - 4 - 5 602 129
M64303.1 - 4 - 5 602 129
S59012.1 - 4 - 5 584 129

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene LGALS3

Panther Classification:

Molecular Function -

extracellular matrix protein

Gene Ontology Categories:

Function(s) Process(es)

monocyte chemotaxis
mRNA processing
RNA splicing
neutrophil chemotaxis
epithelial cell differentiation
regulation of T cell proliferation
innate immune response
negative regulation of endocytosis
eosinophil chemotaxis
macrophage chemotaxis
negative regulation of T cell receptor signaling pathway
positive chemotaxis
regulation of T cell apoptotic process
mononuclear cell migration
positive regulation of mononuclear cell migration
positive regulation of calcium ion import
regulation of extrinsic apoptotic signaling pathway via death domain receptors
negative regulation of immunological synapse formation
negative regulation of T cell activation via T cell receptor contact with antigen bound to MHC molecule on antigen presenting cell
positive regulation of dendritic cell differentiation
negative regulation of extrinsic apoptotic signaling pathway
protein binding
IgE binding
carbohydrate binding
chemoattractant activity
laminin binding
poly(A) RNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0