This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07288623_m1
See other KCNG1 GE Assays ›
Gene Symbol:
KCNG1
Gene Name:
potassium voltage-gated channel modifier subfamily G member 1
Gene Aliases:
K13, KCNG, KV6.1, kH2
Chromosome Location:
Chr.20: 51003656 - 51023176 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs07288623_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: KCNG1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_002237.3 NP_002228.2 1 - 2 272 78
GenBank mRNA AK290689.1 - 1 - 2 273 78
BC046629.2 - 1 - 2 241 5287759 78
DA580190.1 - 1 - 2 279 78
DA684016.1 - 1 - 2 241 78

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK290689.1, BC046629.2, NM_002237.3 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene KCNG1

Panther Classification:

Molecular Function -

voltage-gated ion channel

Gene Ontology Categories:

Function(s) Process(es)

potassium ion transport
protein homooligomerization
potassium ion transmembrane transport
regulation of delayed rectifier potassium channel activity
delayed rectifier potassium channel activity
potassium channel activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0