This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07288512_g1
See other PTGDS GE Assays ›
Gene Symbol:
PTGDS
Gene Name:
prostaglandin D2 synthase
Gene Aliases:
L-PGDS, LPGDS, PDS, PGD2, PGDS, PGDS2
Chromosome Location:
Chr.9: 136977504 - 136981742 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs07288512_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PTGDS
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_000954.5 NP_000945.3 2 - 3 329 64
GenBank mRNA AK075333.1 - 2 - 3 329 64
AK312817.1 - 2 - 3 316 64
AY026356.1 - 2 - 3 254 64
BC005939.1 - 2 - 3 263 4294999 64
BC041463.1 - 4 - 5 1537 64
BG037037.1 - 2 - 3 334 64
BM686612.1 - 2 - 3 317 64
BT019921.1 - 2 - 3 254 64
BT019922.1 - 2 - 3 254 64
M61900.2 - 2 - 3 319 64

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene PTGDS

Panther Classification:

Molecular Function -

transfer/carrier protein

Gene Ontology Categories:

Function(s) Process(es)

prostaglandin biosynthetic process
transport
cyclooxygenase pathway
regulation of circadian sleep/wake cycle, sleep
response to glucocorticoid
negative regulation of male germ cell proliferation
prostaglandin-D synthase activity
transporter activity
retinoid binding
fatty acid binding
protein binding
small molecule binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0