This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07288413_m1
See other ATP6AP1L GE Assays ›
Gene Symbol:
ATP6AP1L
Gene Name:
ATPase H+ transporting accessory protein 1 like
Gene Aliases:
-
Chromosome Location:
Chr.5: 82278645 - 82344294 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs07288413_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ATP6AP1L
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_011543719.2 XP_011542021.1 6 - 7 1490 67
XM_011543720.2 XP_011542022.1 5 - 6 1413 67
XM_011543722.2 XP_011542024.1 6 - 7 1507 67
XM_017010025.1 XP_016865514.1 6 - 7 1136 67
XM_017010026.1 XP_016865515.1 7 - 8 1593 67
XM_017010027.1 XP_016865516.1 7 - 8 941 67
XM_017010028.1 XP_016865517.1 7 - 8 1226 67
XM_017010029.1 XP_016865518.1 6 - 7 1542 67
XM_017010030.1 XP_016865519.1 6 - 7 1175 67
XM_017010031.1 XP_016865520.1 7 - 8 1581 67
XM_017010032.1 XP_016865521.1 8 - 9 1751 67
XM_017010033.1 XP_016865522.1 7 - 8 1214 67
XM_017010034.1 XP_016865523.1 6 - 7 1539 67
XM_017010035.1 XP_016865524.1 5 - 6 1099 67
XM_017010036.1 XP_016865525.1 6 - 7 1514 67
XM_017010037.1 XP_016865526.1 5 - 6 1451 67
XM_017010038.1 XP_016865527.1 6 - 7 1495 67

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

ATP synthase

Gene Ontology Categories:

Function(s) Process(es)

ATP hydrolysis coupled proton transport
proton-transporting ATP synthase activity, rotational mechanism
proton-transporting ATPase activity, rotational mechanism

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7db4fb6694-9hb2s:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header