This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07288079_m1
See other RPGRIP1L GE Assays ›
Gene Symbol:
RPGRIP1L
Gene Name:
RPGRIP1 like
Gene Aliases:
CORS3, FTM, JBTS7, MKS5, NPHP8, PPP1R134
Chromosome Location:
Chr.16: 53599239 - 53703934 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs07288079_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RPGRIP1L
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK304790.1 - 1 - 2 89 84
BX340768.2 - 1 - 2 138 84
CD674885.1 - 1 - 2 59 84
DC423209.1 - 1 - 2 89 84

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK304790.1 transcripts.

Set Membership:

Amplicon length between 71 and 85 Homo sapiens Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0