This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07287963_m1
See other CEBPZOS GE Assays ›
Gene Symbol:
CEBPZOS
Gene Name:
CEBPZ opposite strand
Gene Aliases:
CEBPZ-AS1
Chromosome Location:
Chr.2: 37196451 - 37216193 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs07287963_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CEBPZOS
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001322373.1 NP_001309302.1 4 - 5 392 87
NM_001322374.1 NP_001309303.1 3 - 4 213 87
NM_001322375.1 NP_001309304.1 3 - 4 213 87
NM_001322376.1 NP_001309305.1 3 - 4 213 87
NR_136316.1 3 - 4 213 87
XM_017003106.1 XP_016858595.1 4 - 5 421 87
XM_017003107.1 XP_016858596.1 3 - 4 242 87
XM_017003108.1 XP_016858597.1 3 - 4 223 87
XM_017003109.1 XP_016858598.1 3 - 4 223 87
GenBank mRNA AK001814.1 - 3 - 4 176 87
BC017652.1 - 3 - 4 319 87
BC051746.1 - 2 - 3 162 87
BG109338.1 - 177 87
BM838731.1 - 3 - 4 174 87

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Homo sapiens Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0