This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07287913_m1
See other CAMK2D GE Assays ›
Gene Symbol:
CAMK2D
Gene Name:
calcium/calmodulin dependent protein kinase II delta
Gene Aliases:
CAMKD
Chromosome Location:
Chr.4: 113451032 - 113761927 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs07287913_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CAMK2D
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001321592.1 NP_001308521.1 2 - 3 1014 100

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

Gene Ontology Categories:

Function(s) Process(es)

G1/S transition of mitotic cell cycle
MAPK cascade
regulation of cell growth
response to hypoxia
regulation of the force of heart contraction
regulation of membrane depolarization
regulation of transcription from RNA polymerase II promoter
protein phosphorylation
calcium ion transport
regulation of heart contraction
regulation of G2/M transition of mitotic cell cycle
positive regulation of cardiac muscle hypertrophy
regulation of cell communication by electrical coupling
positive regulation of cardiac muscle cell apoptotic process
regulation of release of sequestered calcium ion into cytosol by sarcoplasmic reticulum
regulation of cardiac muscle contraction by regulation of the release of sequestered calcium ion
positive regulation of smooth muscle cell migration
peptidyl-serine phosphorylation
peptidyl-threonine phosphorylation
cellular potassium ion homeostasis
endoplasmic reticulum calcium ion homeostasis
positive regulation of Rac protein signal transduction
positive regulation of GTPase activity
protein autophosphorylation
positive regulation of smooth muscle cell proliferation
protein oligomerization
relaxation of cardiac muscle
regulation of ryanodine-sensitive calcium-release channel activity
interferon-gamma-mediated signaling pathway
regulation of cellular localization
cell growth involved in cardiac muscle cell development
positive regulation of ERK1 and ERK2 cascade
cellular response to calcium ion
cardiac muscle cell contraction
regulation of heart rate by cardiac conduction
regulation of cardiac muscle cell action potential
regulation of cardiac muscle cell action potential involved in regulation of contraction
regulation of cellular response to heat
regulation of histone deacetylase activity
regulation of cell communication by electrical coupling involved in cardiac conduction
regulation of relaxation of cardiac muscle
negative regulation of sodium ion transmembrane transport
regulation of calcium ion transmembrane transport via high voltage-gated calcium channel
negative regulation of sodium ion transmembrane transporter activity
protein serine/threonine kinase activity
calmodulin-dependent protein kinase activity
Ras guanyl-nucleotide exchange factor activity
protein binding
calmodulin binding
ATP binding
sodium channel inhibitor activity
titin binding
protein homodimerization activity
ion channel binding
nitric-oxide synthase binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0