This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07287815_g1
See other ATP6V1G3 GE Assays ›
Gene Symbol:
ATP6V1G3
Gene Name:
ATPase H+ transporting V1 subunit G3
Gene Aliases:
ATP6G3, Vma10
Chromosome Location:
Chr.1: 198523222 - 198540945 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs07287815_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ATP6V1G3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001320218.1 NP_001307147.1 3 - 4 203 80
NM_133326.1 NP_579872.1 3 - 4 231 108
XM_011509186.2 XP_011507488.1 2 - 3 215 80
XM_011509187.2 XP_011507489.1 2 - 3 256 108
GenBank mRNA BC101129.2 - 2 - 3 121 40014378 80
BC101131.2 - 2 - 3 151 40014383 108
BF509031.1 - 2 - 3 461 108

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene ATP6V1G3

Panther Classification:

Molecular Function -

ATP synthase

Gene Ontology Categories:

Function(s) Process(es)

insulin receptor signaling pathway
transferrin transport
ion transmembrane transport
phagosome acidification
hydrogen ion transmembrane transport
hydrogen-exporting ATPase activity, phosphorylative mechanism
ATPase binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0