This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07287805_m1
See other CRADD GE Assays ›
Gene Symbol:
CRADD
Gene Name:
CASP2 and RIPK1 domain containing adaptor with death domain
Gene Aliases:
MRT34, RAIDD
Chromosome Location:
Chr.12: 93677342 - 93894840 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs07287805_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CRADD
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001320100.1 NP_001307029.1 1 - 2 102 97
NM_001320101.1 NP_001307030.1 1 - 2 102 97
NM_003805.4 NP_003796.1 1 - 2 102 97
NR_135147.1 1 - 2 102 97
XM_017020142.1 XP_016875631.1 1 - 2 131 97
XM_017020143.1 XP_016875632.1 1 - 2 125 97
XM_017020144.1 XP_016875633.1 1 - 2 125 97
XM_017020145.1 XP_016875634.1 1 - 2 124 97
GenBank mRNA AI791547.1 - 1 - 2 96 97
AK294986.1 - 1 - 2 90 97
AK296225.1 - 1 - 2 100 97
AK314861.1 - 1 - 2 82 97
BC017042.1 - 1 - 2 74 3849848 97
HY113325.1 - 1 - 2 103 97
U79115.1 - 1 - 2 94 97

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK294986.1, AK296225.1, AK314861.1, BC017042.1, NM_001320100.1, NM_001320101.1, NM_003805.4, U79115.1, XM_017020142.1, XM_017020143.1, XM_017020144.1, XM_017020145.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene CRADD

Gene Ontology Categories:

Function(s) Process(es)

activation of cysteine-type endopeptidase activity involved in apoptotic process
DNA damage response, signal transduction by p53 class mediator resulting in cell cycle arrest
extrinsic apoptotic signaling pathway via death domain receptors
regulation of apoptotic process
cellular response to mechanical stimulus
positive regulation of apoptotic signaling pathway
protease binding
protein binding
protein binding, bridging
death domain binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0