This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07287420_m1
See other DDX19B GE Assays ›
Gene Symbol:
DDX19B
Gene Name:
DEAD-box helicase 19B
Gene Aliases:
DBP5, DDX19, RNAh
Chromosome Location:
Chr.16: 70289767 - 70335213 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs07287420_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: DDX19B
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001014449.2 NP_001014449.1 4 - 5 532 77
NM_001014451.2 NP_001014451.1 5 - 6 542 77
NM_001257172.1 NP_001244101.1 5 - 6 467 77
NM_001257173.1 NP_001244102.1 5 - 6 624 77
NM_001257174.1 NP_001244103.1 4 - 5 578 77
NM_001257175.1 NP_001244104.1 3 - 4 439 77
NM_007242.5 NP_009173.1 6 - 7 635 77
XM_006721127.2 XP_006721190.1 5 - 6 557 77
XM_011522832.2 XP_011521134.1 7 - 8 691 77
XM_011522833.2 XP_011521135.1 6 - 7 598 77
XM_011522834.2 XP_011521136.1 6 - 7 580 77
XM_017022890.1 XP_016878379.1 6 - 7 673 77
GenBank mRNA AF353720.1 - 4 - 5 578 77
AJ237946.1 - 6 - 7 496 77
AK027378.1 - 6 - 7 600 77
AK223115.1 - 6 - 7 606 77
AK296868.1 - 3 - 4 436 77
AK301938.1 - 5 - 6 624 77
AK302107.1 - 5 - 6 467 77
AK316346.1 - 4 - 5 513 77
AL136639.1 - 5 - 6 511 77
BC003626.2 - 6 - 7 599 3030038 77
BC010008.2 - 4 - 5 484 4298615 77
BC110805.1 - 5 - 6 406 77
BI756088.1 - 2 - 3 293 77
CR457215.1 - 6 - 7 496 77
CR533509.1 - 5 - 6 403 77

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK296868.1, NM_001257175.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene DDX19B

Gene Ontology Categories:

Function(s) Process(es)

mRNA export from nucleus
RNA secondary structure unwinding
protein transport
RNA binding
ATP-dependent RNA helicase activity
helicase activity
protein binding
ATP binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0